accessibility menu, dialog, popup

Test

CAS RN 105-68-0

CAS RN 105-68-0

IUPAC Name: 3-methylbutyl propanoate
Synonyms: isoamyl propionate
Molecular Weight (g/mol): 144.21
Molecular Formula: C8H16O2
InChi Key: XAOGXQMKWQFZEM-UHFFFAOYSA-N
SMILES: CCC(=O)OCCC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results