accessibility menu, dialog, popup

UserName

CAS RN 10489-46-0

RhRhOSOOOOSOOOOSOOO

CAS RN 10489-46-0

IUPAC Name: dirhodium(3+) trisulfate
Synonyms: dirhodium(3+) trisulfate
Molecular Weight (g/mol): 493.98
Molecular Formula: O12Rh2S3
InChi Key: YWFDDXXMOPZFFM-UHFFFAOYSA-H
SMILES: [Rh+3].[Rh+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results