accessibility menu, dialog, popup

UserName

CH3FOCl

CAS RN 104451-99-2

IUPAC Name: 2-chloro-6-fluoro-3-methylbenzaldehyde
Synonyms: 2-chloro-6-fluoro-3-methylbenzaldehyde
Molecular Weight (g/mol): 172.58
Molecular Formula: C8H6ClFO
InChi Key: CSYKEPRGLYWJCW-UHFFFAOYSA-N
SMILES: CC1=CC=C(F)C(C=O)=C1Cl
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)