accessibility menu, dialog, popup

UserName

CAS RN 1044-49-1

IUPAC Name: bis(4-nitrophenyl)-1,3,4-oxadiazole
Synonyms: bis(4-nitrophenyl)-1,3,4-oxadiazole
Molecular Weight (g/mol): 312.24
Molecular Formula: C14H8N4O5
InChi Key: NXVIYWNQRSLRGX-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)C1=NN=C(O1)C1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results