accessibility menu, dialog, popup

Test

CAS RN 104206-65-7

CAS RN 104206-65-7

IUPAC Name: 2-[2-nitro-4-(trifluoromethyl)benzoyl]cyclohexane-1,3-dione
Synonyms: nitisinone nitisinonum nitisinona orfadin
Molecular Weight (g/mol): 329.23
Molecular Formula: C14H10F3NO5
InChi Key: OUBCNLGXQFSTLU-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=CC=C1C(=O)C1C(=O)CCCC1=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Color 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)