CAS RN 104-90-5
CAS RN 104-90-5
IUPAC Name:
5-ethyl-2-methylpyridine
Synonyms:
5-ethyl-2-methylpyridine
Molecular Weight (g/mol):
121.18
Molecular Formula:
C8H11N
InChi Key:
NTSLROIKFLNUIJ-UHFFFAOYSA-N
SMILES:
CCC1=CC=C(C)N=C1
1
–
1
of
1
results
1
5-Ethyl-2-methylpyridine, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.