CAS RN 104-46-1
CAS RN 104-46-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Anethole, MedChemExpress
MedChemExpress Anethole is a type of aromatic compound that occurs widely in nature, widely used as a flavoring substance.