accessibility menu, dialog, popup

UserName

NH2SOHO

CAS RN 104-18-7

IUPAC Name: 2-[(4-aminophenyl)sulfanyl]acetic acid
Synonyms: [(4-aminophenyl)sulfanyl]acetic acid
Molecular Weight (g/mol): 183.23
Molecular Formula: C8H9NO2S
InChi Key: CTPIHHXCACYCIV-UHFFFAOYSA-N
SMILES: NC1=CC=C(SCC(O)=O)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)