CAS RN 10378-22-0
CAS RN 10378-22-0
IUPAC Name:
2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid hydrate trisodium
Synonyms:
edetic acid hydrate trisodium
Molecular Weight (g/mol):
379.23
Molecular Formula:
C10H18N2Na3O9
InChi Key:
DFCJTQJARZUURB-UHFFFAOYSA-N
SMILES:
O.[Na].[Na].[Na].OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
1
–
1
of
1
results