accessibility menu, dialog, popup

UserName

CAS RN 1037-50-9

IUPAC Name: sodium (4-aminophenyl)[(2,6-dimethoxypyrimidin-4-yl)imino]-λ⁶-sulfanoylolate
Synonyms: sodium (4-aminophenyl)[(2,6-dimethoxypyrimidin-4-yl)imino]-λ⁶-sulfanoylolate
Molecular Weight (g/mol): 332.31
Molecular Formula: C12H13N4NaO4S
InChi Key: QEBVWFBANDRUGC-UHFFFAOYSA-M
SMILES: [Na+].COC1=CC(N=S([O-])(=O)C2=CC=C(N)C=C2)=NC(OC)=N1
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results