accessibility menu, dialog, popup

UserName

CAS RN 10351-19-6

IUPAC Name: 2-(pyridin-4-ylsulfanyl)acetate
Synonyms: 2-(pyridin-4-ylsulfanyl)acetate
Molecular Weight (g/mol): 168.19
Molecular Formula: C7H6NO2S
InChi Key: PGUPJAPHYIEKLT-UHFFFAOYSA-M
SMILES: [O-]C(=O)CSC1=CC=NC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results