CAS RN 1034616-18-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Onvansertib, MedChemExpress
MedChemExpress NMS-1286937 is a potent, selective and orally available PLK1 inhibitor, with an IC50 of 2 nM.