CAS RN 1032229-33-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
A939572, MedChemExpress
MedChemExpress A939572 is a potent, and orally bioavailable stearoyl-CoA desaturase1 (SCD1) inhibitor with IC50 values of mSCD1 and hSCD1, respectively.