CAS RN 103177-37-3
IUPAC Name:
N-[4-oxo-2-(2H-1,2,3,4-tetrazol-5-yl)-4H-chromen-7-yl]-4-(4-phenylbutoxy)benzamide
Synonyms:
pranlukast
Molecular Weight (g/mol):
481.51
Molecular Formula:
C27H23N5O4
InChi Key:
UAJUXJSXCLUTNU-UHFFFAOYSA-N
SMILES:
O=C(NC1=CC=C2C(=O)C=C(OC2=C1)C1=NNN=N1)C1=CC=C(OCCCCC2=CC=CC=C2)C=C1
1
–
5
of
5
results
1
Pranlukast, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Pranlukast, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Pranlukast, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Pranlukast, MedChemExpress
MedChemExpress Pranlukast is a highly potent, selective and competitive antagonist of peptide leukotrienes. Pranlukast inhibits [3H]LTE4, [3H]LTD4, and [3H]LTC4 bindings to lung membranes with Kis of 0.63±0.11, 0.99±0.19, and 5640±680 nM, respectively.