accessibility menu, dialog, popup

UserName

CAS RN 10311-84-9

H3COPSOCH3SClNOO

CAS RN 10311-84-9

IUPAC Name: O,O-diethyl {[2-chloro-1-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)ethyl]sulfanyl}phosphonothioate
Synonyms: dialifor
Molecular Weight (g/mol): 393.84
Molecular Formula: C14H17ClNO4PS2
InChi Key: MUMQYXACQUZOFP-UHFFFAOYNA-N
SMILES: CCOP(=S)(OCC)SC(CCl)N1C(=O)C2=CC=CC=C2C1=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results