CAS RN 10304-16-2
CAS RN 10304-16-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Thietan-3-ol, TRC
CAS: 10304-16-2 Molecular Formula: C3H6OS Molecular Weight (g/mol): 90.14 IUPAC Name: thietan-3-ol SMILES: OC1CSC1