CAS RN 1030017-51-6
CAS RN 1030017-51-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
L 888607 Racemate, MedChemExpress
MedChemExpress L 888607 Racemate is a selective prostaglandin D2 receptor subtype 1 (DP1) antagonist, with Kis of 132 nM and 17 nM for DP1 and thromboxane A2 receptor (TP), respectively.
