CAS RN 1029439-85-7
CAS RN 1029439-85-7
IUPAC Name:
2,3-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Synonyms:
2,3-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Molecular Weight (g/mol):
256.06
Molecular Formula:
C12H15BF2O3
InChi Key:
JNBBZUYJDRTJCN-UHFFFAOYSA-N
SMILES:
CC1(C)OB(OC1(C)C)C1=C(F)C(F)=C(O)C=C1
1
–
1
of
1
results
1
2,3-Difluoro-4-hydroxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific™
CAS: 1029439-85-7 Molecular Formula: C12H15BF2O3 Molecular Weight (g/mol): 256.06 MDL Number: MFCD22414554 InChI Key: JNBBZUYJDRTJCN-UHFFFAOYSA-N Synonym: 2,3-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol,2,3-difluoro-4-hydroxyphenylboronic acid pinacol ester PubChem CID: 57655377 IUPAC Name: 2,3-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol SMILES: CC1(C)OB(OC1(C)C)C1=C(F)C(F)=C(O)C=C1