CAS RN 10294-38-9
IUPAC Name:
barium(2+) hydrate dichlorate
Synonyms:
barium(2+) hydrate dichlorate
Molecular Weight (g/mol):
322.24
Molecular Formula:
BaCl2H2O7
InChi Key:
CIADYQPMTWAHHH-UHFFFAOYSA-L
SMILES:
O.[Ba++].[O-][Cl](=O)=O.[O-][Cl](=O)=O
1
–
1
of
1
results
1
Alfa Aesar™ Barium chlorate monohydrate, 98%, Thermo Scientific™
CAS: 10294-38-9 Molecular Formula: BaCl2H2O7 Molecular Weight (g/mol): 322.236 MDL Number: MFCD00149147 InChI Key: CIADYQPMTWAHHH-UHFFFAOYSA-L Synonym: barium chlorate monohydrate,barium 2+ dichlorate hydrate,ba.2clo3.h2o,chloric acid, bariumsalt 2:1,barium 2+ hydrate dichlorate PubChem CID: 6096944 IUPAC Name: barium(2+);dichlorate;hydrate SMILES: O.[O-]Cl(=O)=O.[O-]Cl(=O)=O.[Ba+2]