CAS RN 10293-06-8
CAS RN 10293-06-8
IUPAC Name:
3-bromo-1,7,7-trimethylbicyclo[2.2.1]heptan-2-one
Synonyms:
3-bromo-d-camphor
Molecular Weight (g/mol):
231.13
Molecular Formula:
C10H15BrO
InChi Key:
NJQADTYRAYFBJN-UHFFFAOYNA-N
SMILES:
CC1(C)C2CCC1(C)C(=O)C2Br
1
–
3
of
3
results
1
(+)-3-Bromocamphor, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(+)-3-Bromocamphor, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(+)-3-Bromocamphor, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.