CAS RN 102804-82-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
4-Desmethoxypropoxyl-4-methoxy S-Deoxo Rabeprazole, TRC
CAS: 102804-82-0 Molecular Formula: C15 H15 N3 O S Molecular Weight (g/mol): 285.36 Synonym: 2-[[(4-Methoxy-3-methyl-2-pyridinyl)methyl]thio]-1H-benzimidazole,Rabeprazole Sodium Imp G (EP) IUPAC Name: 2-[(4-methoxy-3-methylpyridin-2-yl)methylsulfanyl]-1H-benzimidazole SMILES: COc1ccnc(CSc2nc3ccccc3[nH]2)c1C
2
2-[[(4-Methoxy-3-methylpyridin-2-yl)methyl]sulfanyl]-1H-benzimidazole, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.