CAS RN 1027011-64-8
CAS RN 1027011-64-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
(6Alpha,9Beta,11Beta)-9,11-Epoxy-6-fluoropregna-1,4,16-triene-3,20-dione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(6Alpha,9Beta,11Beta)-9,11-Epoxy-6-fluoropregna-1,4,16-triene-3,20-dione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.