Learn More
(6Alpha,9Beta,11Beta)-9,11-Epoxy-6-fluoropregna-1,4,16-triene-3,20-dione, TRC
Specifications
Specifications
| Chemical Name or Material | (6alpha,9beta,11beta)-9,11-Epoxy-6-fluoropregna-1,4,16-triene-3,20-dione |
| CAS | 1027011-64-8 |
| Molecular Formula | C21H23FO3 |
| Quantity | 2.5 mg |
| InChI Formula | InChI=1S/C21H23FO3/c1-11(23)13-4-5-14-15-9-17(22)16-8-12(24)6-7-20(16,3)21(15)18(25-21)10-19(13,14)2/h4,6-8,14-15,17-18H,5,9-10H2,1-3H3/t14-,15?,17-,18+,19-,20+,21-/m1/s1 |
| SMILES | CC(=O)C1=CC[C@@H]2C3C[C@@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]35O[C@H]5C[C@]12C |
| Molecular Weight (g/mol) | 342.4 |
| Formula Weight | 342.1631 |