accessibility menu, dialog, popup

UserName

CAS RN 102649-78-5

IUPAC Name: 5-chloro-N-(6-phenylhexyl)naphthalene-1-sulfonamide
Synonyms: 5-chloro-N-(6-phenylhexyl)naphthalene-1-sulfonamide
Molecular Weight (g/mol): 401.95
Molecular Formula: C22H24ClNO2S
InChi Key: YUXMTDYHFPRPLG-UHFFFAOYSA-N
SMILES: ClC1=C2C=CC=C(C2=CC=C1)S(=O)(=O)NCCCCCCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)