accessibility menu, dialog, popup

UserName

CAS RN 102587-98-4

IUPAC Name: 2,7-dibromopyrene
Synonyms: 2,7-dibromopyrene
Molecular Weight (g/mol): 360.05
Molecular Formula: C16H8Br2
InChi Key: IGTQPXMEWQTTBJ-UHFFFAOYSA-N
SMILES: BrC1=CC2=CC=C3C=C(Br)C=C4C=CC(=C1)C2=C34
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)