CAS RN 102491-80-5
CAS RN 102491-80-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Demethyl Colchicine, TRC
CAS: 102491-80-5 Molecular Formula: C21 H23 N O6 Molecular Weight (g/mol): 385.41 IUPAC Name: N-[(7S)-2-hydroxy-1,3,10-trimethoxy-9-oxo-6,7-dihydro-5H-benzo[a]heptalen-7-yl]acetamide SMILES: COC1=CC=C2C(=CC1=O)[C@H](CCc3cc(OC)c(O)c(OC)c23)NC(=O)C