CAS RN 102394-28-5
CAS RN 102394-28-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
G 7460, TRC
CAS: 102394-28-5 Molecular Formula: C14 H15 N5 O7 S Molecular Weight (g/mol): 397.36 Synonym: Benzoic acid, 4-hydroxy-2-[[[[(4-methoxy-6-methyl-1,3,5-triazin-2-yl)amino]carbonyl]amino]sulfonyl]-, methyl ester,G 7460 IUPAC Name: methyl 4-hydroxy-2-[(4-methoxy-6-methyl-1,3,5-triazin-2-yl)carbamoylsulfamoyl]benzoate SMILES: COC(=O)c1ccc(O)cc1S(=O)(=O)NC(=O)Nc2nc(C)nc(OC)n2