CAS RN 1023805-47-1
IUPAC Name:
N-(2,6-difluorophenyl)-3-nitrobenzene-1-sulfonamide
Synonyms:
N-(2,6-difluorophenyl)-3-nitrobenzenesulfonamide
Molecular Weight (g/mol):
314.26
Molecular Formula:
C12H8F2N2O4S
InChi Key:
MTBZLMOKCNKJCR-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC(=CC=C1)S(=O)(=O)NC1=C(F)C=CC=C1F
1
–
1
of
1
results
1
N-(2,6-Difluorophenyl)-3-nitrobenzenesulfonamide, 97%, Thermo Scientific™
CAS: 1023805-47-1 Molecular Formula: C12H8F2N2O4S Molecular Weight (g/mol): 314.26 MDL Number: MFCD05815404 InChI Key: MTBZLMOKCNKJCR-UHFFFAOYSA-N Synonym: n-2,6-difluorophenyl-3-nitrobenzenesulfonamide,2,6-difluorophenyl 3-nitrophenyl sulfonyl amine PubChem CID: 4288877 IUPAC Name: N-(2,6-difluorophenyl)-3-nitrobenzenesulfonamide SMILES: [O-][N+](=O)C1=CC(=CC=C1)S(=O)(=O)NC1=C(F)C=CC=C1F