CAS RN 102342-85-8
CAS RN 102342-85-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Malonic Acid-1,2,3-13C3, TRC
CAS: 102342-85-8 Molecular Formula: 13C3H4O4 Molecular Weight (g/mol): 107.04 Synonym: Propanedioic Acid-1,2,3-13C3,1,3-Propanedioic Acid-1,2,3-13C3,Carboxyacetic Acid-1,2,3-13C3,Dicarboxymethane-1,2,3-13C3,Methanedicarboxylic Acid-1,2,3-13C3,NSC 8124-2-1,2,3-13C3,[1,2,3-13C3]-Malonic acid SMILES: O[13C]([13C]([H])([H])[13C](O)=O)=O