CAS RN 10222-01-2
CAS RN 10222-01-2
IUPAC Name:
2,2-dibromo-2-cyanoacetamide
Synonyms:
dbnpa
Molecular Weight (g/mol):
241.87
Molecular Formula:
C3H2Br2N2O
InChi Key:
UUIVKBHZENILKB-UHFFFAOYSA-N
SMILES:
NC(=O)C(Br)(Br)C#N
1
–
1
of
1
results
1
Dibromocyanoacetamide, TRC
CAS: 10222-01-2 Molecular Formula: C3 H2 Br2 N2 O Molecular Weight (g/mol): 241.87 Synonym: 2,2-Dibromo-2-cyanoacetamide,2,2-Dibromo-3-nitrilopropanamide,2,2-Dibromo-3-nitrilopropionamide,2-Cyano-2,2-dibromoacetamide,AM 7287,Amerstat 300,BE 3S,Biomate 723,Busan 94,DB 20,DBNPA,DBNPA 7287,Dibromocyanoacetamide,Dibromonitrilopropionamide,Dow antimicrobial 7287,Dow bactericide 7287,Dowicil QK 20,Mucosin NT,NSC 98283,Odycide B 340,QK 20,Thor DB 20,X-CIDE 508,X-CIDE 509 IUPAC Name: 2,2-dibromo-2-cyanoacetamide SMILES: NC(=O)C(Br)(Br)C#N