CAS RN 1021343-21-4
CAS RN 1021343-21-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-(Nonylthio)propanamide, TRC
CAS: 1021343-21-4 Molecular Formula: C6 H10 O2 Molecular Weight (g/mol): 114.142 IUPAC Name: 1-methylcyclobutane-1-carboxylic acid SMILES: CC1(CCC1)C(=O)O