accessibility menu, dialog, popup

UserName

CAS RN 10210-32-9

H3CO

CAS RN 10210-32-9

IUPAC Name: 1-(anthracen-2-yl)ethan-1-one
Synonyms: 1-(anthracen-2-yl)ethanone
Molecular Weight (g/mol): 220.27
Molecular Formula: C16H12O
InChi Key: DQFWGPPCMONVDS-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C2C=C3C=CC=CC3=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results