accessibility menu, dialog, popup

UserName

OOFF(E)

CAS RN 102082-89-3

IUPAC Name: (2E)-3-(2,6-difluorophenyl)prop-2-enoate
Synonyms: (2E)-3-(2,6-difluorophenyl)prop-2-enoate
Molecular Weight (g/mol): 183.13
Molecular Formula: C9H5F2O2
InChi Key: JMUOYANNVIFGFN-SNAWJCMRSA-M
SMILES: [O-]C(=O)\C=C\C1=C(F)C=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results