CAS RN 1020719-54-3
CAS RN 1020719-54-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
9-[2-(Hydroxypropyl-d6] Adenine, TRC
CAS: 1020719-54-3 Molecular Formula: C8 2H6 H5 N5 O Molecular Weight (g/mol): 199.243 Synonym: rac-Desphosphoryl Tenofovir-d6,DL-9-(2-Hydroxypropyl)adenine-d6,6-Amino-α-(methyl-d3)-9H-purine-9-ethan-α,β,β-d3-ol IUPAC Name: 1-(6-aminopurin-9-yl)-1,1,2,3,3,3-hexadeuteriopropan-2-ol SMILES: [2H]C([2H])([2H])C([2H])(O)C([2H])([2H])n1cnc2c(N)ncnc12