CAS RN 102-27-2
CAS RN 102-27-2
IUPAC Name:
N-ethyl-3-methylaniline
Synonyms:
benzenamine, N-ethyl-3-methyl-
Molecular Weight (g/mol):
135.21
Molecular Formula:
C9H13N
InChi Key:
GUYMMHOQXYZMJQ-UHFFFAOYSA-N
SMILES:
CCNC1=CC=CC(C)=C1
1
–
3
of
3
results
1
N-Ethyl-m-toluidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-Ethyl-m-toluidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-Ethyl-m-toluidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.