CAS RN 102-02-3
CAS RN 102-02-3
IUPAC Name:
hydrogen N-(diaminomethylidene)-N''-phenylguanidine chloride
Synonyms:
phenylbiguanide
Molecular Weight (g/mol):
213.67
Molecular Formula:
C8H12ClN5
InChi Key:
FHUDRDSKZQDCBC-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].NC(N)=NC(N)=NC1=CC=CC=C1
1
–
1
of
1
results
1
Medchem Express Phenylbiguanide, MedChemExpress
MedChemExpress Phenylbiguanide is a 5-HT3 receptor selective agonist with an EC50 of 3.0±0.1 μM.
