CAS RN 10184-70-0
CAS RN 10184-70-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
21-Desacetyl Anecortave, TRC
CAS: 10184-70-0 Molecular Formula: C21 H28 O4 Molecular Weight (g/mol): 344.44 Synonym: Pregna-4,9(11)-diene-3,20-dione, 17,21-dihydroxy-,17,21-Dihydroxypregna-4,9(11)-diene-3,20-dione,AL 4940 IUPAC Name: (8S,10S,13S,14S,17R)-17-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-2,6,7,8,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthren-3-one SMILES: C[C@]12CC=C3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@]2(O)C(=O)CO