accessibility menu, dialog, popup

UserName

CAS RN 1017778-27-6

CH3FFFOOH

CAS RN 1017778-27-6

IUPAC Name: 2-[5-methyl-2-(trifluoromethyl)phenyl]acetic acid
Synonyms: [5-methyl-2-(trifluoromethyl)phenyl]acetic acid
Molecular Weight (g/mol): 218.18
Molecular Formula: C10H9F3O2
InChi Key: UMEQVNCETMDPES-UHFFFAOYSA-N
SMILES: CC1=CC(CC(O)=O)=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results