accessibility menu, dialog, popup

UserName

CAS RN 1017777-48-8

H3COClClF

CAS RN 1017777-48-8

IUPAC Name: 1,2-dichloro-4-fluoro-3-methoxybenzene
Synonyms: 1,2-dichloro-4-fluoro-3-methoxybenzene
Molecular Weight (g/mol): 195.01
Molecular Formula: C7H5Cl2FO
InChi Key: MXQUAHMSEJQMOE-UHFFFAOYSA-N
SMILES: COC1=C(F)C=CC(Cl)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results