accessibility menu, dialog, popup

UserName

NaNaOPOFO

CAS RN 10163-15-2

IUPAC Name: disodium phosphorofluoridate
Synonyms: sodium monofluorophosphate
Molecular Weight (g/mol): 143.95
Molecular Formula: FNa2O3P
InChi Key: BFDWBSRJQZPEEB-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]P([O-])(F)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)