accessibility menu, dialog, popup

UserName

CaCaOSiOOO

CAS RN 10101-39-0

IUPAC Name: dicalcium silicate
Synonyms: calcium silicate
Molecular Weight (g/mol): 172.24
Molecular Formula: Ca2O4Si
InChi Key: JHLNERQLKQQLRZ-UHFFFAOYSA-N
SMILES: [Ca++].[Ca++].[O-][Si]([O-])([O-])[O-]
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)