accessibility menu, dialog, popup

UserName

H3CHNONOONOO

CAS RN 10056-18-5

IUPAC Name: 3,5-dinitro-N-propylbenzamide
Synonyms: 3,5-dinitro-N-propylbenzamide
Molecular Weight (g/mol): 253.21
Molecular Formula: C10H11N3O5
InChi Key: LTJACLQPFWDXMR-UHFFFAOYSA-N
SMILES: CCCNC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results