accessibility menu, dialog, popup

UserName

NNOBr

CAS RN 1004550-20-2

IUPAC Name: 3-bromoimidazo[1,2-a]pyridine-6-carbaldehyde
Synonyms: 3-bromoimidazo[1,2-a]pyridine-6-carbaldehyde
Molecular Weight (g/mol): 225.05
Molecular Formula: C8H5BrN2O
InChi Key: PMCPYYAVCSERGH-UHFFFAOYSA-N
SMILES: BrC1=CN=C2C=CC(C=O)=CN12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results