accessibility menu, dialog, popup

UserName

BNClCH3HOOH

CAS RN 1003043-40-0

IUPAC Name: (6-chloro-5-methylpyridin-3-yl)boronic acid
Synonyms: 6-chloro-5-methylpyridin-3-ylboronic acid
Molecular Weight (g/mol): 171.39
Molecular Formula: C6H7BClNO2
InChi Key: WWOOKOBRTKSRLV-UHFFFAOYSA-N
SMILES: CC1=C(Cl)N=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results