accessibility menu, dialog, popup

UserName

CAS RN 1000932-18-2

OHOOFF

CAS RN 1000932-18-2

IUPAC Name: 3-[4-(difluoromethoxy)phenyl]propanoic acid
Synonyms: 3-[4-(difluoromethoxy)phenyl]propanoic acid
Molecular Weight (g/mol): 216.18
Molecular Formula: C10H10F2O3
InChi Key: FOBBJWXKCGHQLT-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC=C(OC(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results