accessibility menu, dialog, popup

UserName

H3CONFFF

CAS RN 1000555-32-7

IUPAC Name: 2-[4-methoxy-3-(trifluoromethyl)phenyl]acetonitrile
Synonyms: 2-[4-methoxy-3-(trifluoromethyl)phenyl]acetonitrile
Molecular Weight (g/mol): 215.18
Molecular Formula: C10H8F3NO
InChi Key: DJRVULCEMUTQLB-UHFFFAOYSA-N
SMILES: COC1=C(C=C(CC#N)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results