accessibility menu, dialog, popup

UserName

NClFFF

CAS RN 1000530-58-4

IUPAC Name: 2-[3-chloro-4-(trifluoromethyl)phenyl]acetonitrile
Synonyms: 2-[3-chloro-4-(trifluoromethyl)phenyl]acetonitrile
Molecular Weight (g/mol): 219.59
Molecular Formula: C9H5ClF3N
InChi Key: YJHNYZHRYHJFFK-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C(Cl)C=C(CC#N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results