accessibility menu, dialog, popup

UserName

CH3OOFFF

CAS RN 1000339-55-8

IUPAC Name: 3-methyl-5-(trifluoromethoxy)benzaldehyde
Synonyms: 3-methyl-5-(trifluoromethoxy)benzaldehyde
Molecular Weight (g/mol): 204.15
Molecular Formula: C9H7F3O2
InChi Key: LAGVGNCAIJDRHK-UHFFFAOYSA-N
SMILES: CC1=CC(OC(F)(F)F)=CC(C=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results