Learn More
4-Methyl-3-nitrophenol, 98%
Product Code. 11309828
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
250 g
Unit Size:
10g
250g
2g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11309828 50 g 50g
11319828 2 g 2g
11399818 10 g 10g
11329828 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11309828 Supplier Thermo Scientific Alfa Aesar Supplier No. 044285.18
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
CAS: 2042-14-0 | C7H7NO3 | 153.137 g/mol
It is used as a chemical, pharmaceutical and OLED intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2042-14-0
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00007244
InChI Key BQEXDUKMTVYBRK-UHFFFAOYSA-N
Synonym 3-nitro-4-methylphenol, 3-nitro-p-cresol, 4-methyl-3-nitro-phenol, 4-hydroxy-2-nitrotoluene, 2-nitro-4-hydroxytoluene, phenol, 4-methyl-3-nitro, p-cresol, 3-nitro, unii-4b422hud6h, 4-methyl-5-nitrophenol
PubChem CID 16271
IUPAC Name 4-methyl-3-nitrophenol
SMILES CC1=C(C=C(C=C1)O)[N+](=O)[O-]
Specifications
Melting Point 77°C to 81°C
Odor Phenol-like
Quantity 50 g
UN Number UN2446
Solubility Information Slightly soluble in water.
Formula Weight 153.14
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Methyl-3-nitrophenol