missing translation for 'onlineSavingsMsg'
Learn More

Zosuquidar Trihydrochloride, TRC

Product Code. 30452099
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
Product Code. Quantity unitSize
30452099 10 mg 10mg
30452111 25 mg 25mg
30456972 50 mg 50mg
3 options
This item is not returnable. View return policy

Product Code. 30452099

Brand: Toronto Research Chemicals TRCZ70087010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 167465-36-3
Formula Weight 635.1685
InChI Formula InChI=1S/C32H31F2N3O2.3ClH/c33-32(34)29-22-7-1-3-9-24(22)31(25-10-4-2-8-23(25)30(29)32)37-17-15-36(16-18-37)19-21(38)20-39-28-13-5-12-27-26(28)11-6-14-35-27;;;/h1-14,21,29-31,38H,15-20H2;3*1H/t21-,29-,30+,31-;;;/m1.../s1
Molecular Formula C32 H31 F2 N3 O2 . 3 Cl H
Molecular Weight (g/mol) 636.99
SMILES Cl.Cl.Cl.O[C@@H](COc1cccc2ncccc12)CN3CCN(CC3)[C@H]4c5ccccc5[C@H]6[C@@H](c7ccccc47)C6(F)F
Synonym (αR)-4-[(1aα,6α,10bα)-1,1-Difluoro-1,1a,6,10b-tetrahydrodibenzo[a,e]cyclopropa[c]cyclohepten-6-yl]-α-[(5-quinolinyloxy)methyl]-1-piperazineethanol Hydrochloride, Zosuquidar Hydrochloride, LY 335979, RS 33295-198
Chemical Name or Material Zosuquidar trihydrochloride

Specifications

Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.